Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 25 IgM
SDP
Catalog No. 5000078292
Encompass
The V-PLEX SARS-CoV-2 Panel 2 0 is an immunoassay kit from Meso Scale Diagnostics that detects IgM antibodies to SARS-CoV-2 in human serum and plasma It contains a 96-well plate coated with SARS-CoV-2 antigens detection antibodies calibrators controls diluent and wash buffer The electrochemiluminescence immunoassay takes 3 5 hours to run and has a dynamic range of 50-100 000 pg/mL This semi-quantitative assay aids in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection
Catalog No. 50-000-78292 Supplier Meso Scale Discovery Supplier No. K15584U4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX SARS-CoV-2 Panel 2 0 is an immunoassay kit from Meso Scale Diagnostics that detects IgM antibodies to SARS-CoV-2 in human serum and plasma It contains a 96-well plate coated with SARS-CoV-2 antigens detection antibodies calibrators controls diluent and wash buffer The electrochemiluminescence immunoassay takes 3 5 hours to run and has a dynamic range of 50-100 000 pg/mL This semi-quantitative assay aids in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection