Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 25 MSE IgA
SDP
Catalog No. 5000081443
Encompass
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 2 Mouse IgA Kit is designed to measure IgA antibodies against SARS-CoV-2 in mouse serum and plasma samples This electrochemiluminescence immunoassay allows for the qualitative detection of antibodies targeting the receptor binding domain and nucleoprotein of SARS-CoV-2 The kit includes a 96-well plate coated with antigens detection antibodies calibrators controls diluent read buffer and wash buffer sufficient for 25 tests in duplicate when using the recommended sample dilution This assay can aid in the evaluation of vaccine candidates or therapeutics targeting SARS-CoV-2 in mouse models
Catalog No. 50-000-81443 Supplier Meso Scale Discovery Supplier No. K15589U2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 2 Mouse IgA Kit is designed to measure IgA antibodies against SARS-CoV-2 in mouse serum and plasma samples This electrochemiluminescence immunoassay allows for the qualitative detection of antibodies targeting the receptor binding domain and nucleoprotein of SARS-CoV-2 The kit includes a 96-well plate coated with antigens detection antibodies calibrators controls diluent read buffer and wash buffer sufficient for 25 tests in duplicate when using the recommended sample dilution This assay can aid in the evaluation of vaccine candidates or therapeutics targeting SARS-CoV-2 in mouse models