Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 25 MSE IgG
SDP
Catalog No. 5000082453
Encompass
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 2 Mouse IgG Kit is an electrochemiluminescence immunoassay that detects IgG antibodies to SARS-CoV-2 in mouse serum and plasma samples The assay uses a multiplex format to measure antibodies specific for the SARS-CoV-2 spike and nucleocapsid proteins The kit includes a plate coated with antigens detection antibodies calibrators controls diluent and wash buffer sufficient for 25 tests in a 96-well plate format The assay has high sensitivity and specificity for detecting the IgG response to SARS-CoV-2 infection or vaccination in mice
Catalog No. 50-000-82453 Supplier Meso Scale Discovery Supplier No. K15587U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 2 Mouse IgG Kit is an electrochemiluminescence immunoassay that detects IgG antibodies to SARS-CoV-2 in mouse serum and plasma samples The assay uses a multiplex format to measure antibodies specific for the SARS-CoV-2 spike and nucleocapsid proteins The kit includes a plate coated with antigens detection antibodies calibrators controls diluent and wash buffer sufficient for 25 tests in a 96-well plate format The assay has high sensitivity and specificity for detecting the IgG response to SARS-CoV-2 infection or vaccination in mice