Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 25 MSE IgM
SDP
Catalog No. 5000081409
Encompass
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 2 Mouse IgM Kit is an electrochemiluminescence immunoassay that detects IgM antibodies to SARS-CoV-2 in mouse serum and plasma samples The kit includes a 96-well plate coated with SARS-CoV-2 antigens detection antibodies calibrators controls diluent wash buffer read buffer and plate seals The assay has a testing time of approximately 3 5 hours and a dynamic range of 48 4-50000 pg/mL This product can aid in evaluating vaccine efficacy and immunogenicity in mouse models of COVID-19
Catalog No. 50-000-81409 Supplier Meso Scale Discovery Supplier No. K15588U2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 2 Mouse IgM Kit is an electrochemiluminescence immunoassay that detects IgM antibodies to SARS-CoV-2 in mouse serum and plasma samples The kit includes a 96-well plate coated with SARS-CoV-2 antigens detection antibodies calibrators controls diluent wash buffer read buffer and plate seals The assay has a testing time of approximately 3 5 hours and a dynamic range of 48 4-50000 pg/mL This product can aid in evaluating vaccine efficacy and immunogenicity in mouse models of COVID-19