Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 26 ACE2
SDP
Catalog No. 5000082378
Encompass
The Mesoscale K15596U-4 is a 4-channel 16-bit 250 kS/s simultaneous sampling data acquisition module It has 4 analog voltage input channels with programmable input ranges of 10 V 5 V 2 V and 1 V The module has 16-bit resolution 250 kS/s maximum aggregate sample rate and includes software for streaming data to PC over USB 2 0 The compact module is bus-powered and integrates analog input A/D conversion and USB 2 0 communication on a single board It is compatible with both Windows and Linux The K15596U-4 is well-suited for high-accuracy voltage measurement and data logging applications
Catalog No. 50-000-82378 Supplier Meso Scale Discovery Supplier No. K15596U4
Add to Cart
Add to Cart
Request a Quote
The Mesoscale K15596U-4 is a 4-channel 16-bit 250 kS/s simultaneous sampling data acquisition module It has 4 analog voltage input channels with programmable input ranges of 10 V 5 V 2 V and 1 V The module has 16-bit resolution 250 kS/s maximum aggregate sample rate and includes software for streaming data to PC over USB 2 0 The compact module is bus-powered and integrates analog input A/D conversion and USB 2 0 communication on a single board It is compatible with both Windows and Linux The K15596U-4 is well-suited for high-accuracy voltage measurement and data logging applications