Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 26 IgA Kit
SDP
Catalog No. 5000080250
Encompass
The Meso Scale Diagnostics SARS-CoV-2 Panel 2 0 IgA is an in vitro diagnostic test for the qualitative detection of IgA antibodies to SARS-CoV-2 in human serum and plasma This electrochemiluminescence immunoassay utilizes recombinant SARS-CoV-2 antigens to detect IgA antibodies that bind to the receptor binding domain and nucleocapsid protein The test is intended for use as an aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection The panel contains a 96-well plate and all necessary reagents for testing Results are available in under 3 hours This product requires the MSD reader and associated software for analysis
Catalog No. 50-000-80250 Supplier Meso Scale Discovery Supplier No. K15595U2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Diagnostics SARS-CoV-2 Panel 2 0 IgA is an in vitro diagnostic test for the qualitative detection of IgA antibodies to SARS-CoV-2 in human serum and plasma This electrochemiluminescence immunoassay utilizes recombinant SARS-CoV-2 antigens to detect IgA antibodies that bind to the receptor binding domain and nucleocapsid protein The test is intended for use as an aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection The panel contains a 96-well plate and all necessary reagents for testing Results are available in under 3 hours This product requires the MSD reader and associated software for analysis