Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 26 IgG Kit
SDP
Catalog No. 5000081416
Encompass
The Mesoscale K15593U-2 is a high-performance thermoelectric cooler for photonics and semiconductor applications offering precise temperature control It has a 15W heat pumping capacity and a 70 C temperature differential The compact design measures 30 x 30 x 3 4mm requiring a 2 7-3 6V DC input and up to 5A current It includes a thermistor for temperature sensing Key features are low thermal resistance high reliability and RoHS compliance Ideal for stabilizing laser diode temperatures the K15593U-2 delivers robust performance in a small footprint
Catalog No. 50-000-81416 Supplier Meso Scale Discovery Supplier No. K15593U2
Add to Cart
Add to Cart
Request a Quote
The Mesoscale K15593U-2 is a high-performance thermoelectric cooler for photonics and semiconductor applications offering precise temperature control It has a 15W heat pumping capacity and a 70 C temperature differential The compact design measures 30 x 30 x 3 4mm requiring a 2 7-3 6V DC input and up to 5A current It includes a thermistor for temperature sensing Key features are low thermal resistance high reliability and RoHS compliance Ideal for stabilizing laser diode temperatures the K15593U-2 delivers robust performance in a small footprint