Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 26 MSE IgA
SDP
Catalog No. 5000081466
Encompass
The Meso Scale Discovery COVID-19 Spike Protein Neutralization Assay Kit is a high-performance electrochemiluminescent immunoassay that quantitatively measures neutralizing antibodies against SARS-CoV-2 in serum and plasma samples The assay uses recombinant SARS-CoV-2 spike protein coated onto plates to capture antibodies which are detected using an anti-human IgG secondary antibody conjugated to an electrochemiluminescent label The kit includes coated plates detection antibody calibrators controls buffers substrate and plate seals The assay has high sensitivity and a broad dynamic range allowing quantification of antibody levels It is for research use only
Catalog No. 50-000-81466 Supplier Meso Scale Discovery Supplier No. K15599U2
Please call customer service for current pricing. 1-800-766-7000
The Meso Scale Discovery COVID-19 Spike Protein Neutralization Assay Kit is a high-performance electrochemiluminescent immunoassay that quantitatively measures neutralizing antibodies against SARS-CoV-2 in serum and plasma samples The assay uses recombinant SARS-CoV-2 spike protein coated onto plates to capture antibodies which are detected using an anti-human IgG secondary antibody conjugated to an electrochemiluminescent label The kit includes coated plates detection antibody calibrators controls buffers substrate and plate seals The assay has high sensitivity and a broad dynamic range allowing quantification of antibody levels It is for research use only