Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 27 ACE2 Kit
SDP
Catalog No. 5000080931
Encompass
The V-PLEX SARS-CoV-2 Panel 27 (ACE2) Kit (5 PL) K15609U-2 is a multiplex immunoassay kit for the qualitative detection of antibodies to SARS-CoV-2 in human serum and plasma It contains reagents for 5 plates to run up to 27 samples The assay uses recombinant SARS-CoV-2 receptor binding domain (RBD) and spike protein S1 antigens to detect IgG IgA and IgM antibodies and provides a semi-quantitative result This can aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection The kit has high sensitivity and specificity and takes approximately 3 hours to run
Catalog No. 50-000-80931 Supplier Meso Scale Discovery Supplier No. K15609U2
Add to Cart
Add to Cart
Request a Quote
The V-PLEX SARS-CoV-2 Panel 27 (ACE2) Kit (5 PL) K15609U-2 is a multiplex immunoassay kit for the qualitative detection of antibodies to SARS-CoV-2 in human serum and plasma It contains reagents for 5 plates to run up to 27 samples The assay uses recombinant SARS-CoV-2 receptor binding domain (RBD) and spike protein S1 antigens to detect IgG IgA and IgM antibodies and provides a semi-quantitative result This can aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection The kit has high sensitivity and specificity and takes approximately 3 hours to run