Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 27 ACE2
SDP
Catalog No. 5000080881
Encompass
The Meso Scale Discovery K15609U-4 is a 96-well plate that is coated with a proprietary carbon electrode sensor to enable electrochemiluminescent detection of biomarkers It is designed for quantitative measurement of human IL-17A/F heterodimer in cell culture supernatants serum plasma and other biological matrices with a lower limit of detection of 0 04 pg/mL The plate has high sensitivity and broad dynamic range while using a small sample volume of 50 L It is compatible with the MSD QuickPlex SQ 120 instrument and provides rapid results in 3 5 hours The K15609U-4 kit includes a plate detection antibody calibrator and buffer
Catalog No. 50-000-80881 Supplier Meso Scale Discovery Supplier No. K15609U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery K15609U-4 is a 96-well plate that is coated with a proprietary carbon electrode sensor to enable electrochemiluminescent detection of biomarkers It is designed for quantitative measurement of human IL-17A/F heterodimer in cell culture supernatants serum plasma and other biological matrices with a lower limit of detection of 0 04 pg/mL The plate has high sensitivity and broad dynamic range while using a small sample volume of 50 L It is compatible with the MSD QuickPlex SQ 120 instrument and provides rapid results in 3 5 hours The K15609U-4 kit includes a plate detection antibody calibrator and buffer