Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 27 IgA Kit
SDP
Catalog No. 5000080919
Encompass
The V-PLEX Human Cytokine 30-plex Kit is an immunoassay for measuring 30 human cytokines and chemokines simultaneously It uses electrochemiluminescence detection on the MSD platform The kit includes a plate with capture antibodies detection antibodies and calibrator diluents The assay takes about 3 5 hours and works with serum plasma culture supernatants and other samples It offers high sensitivity a broad dynamic range and low sample volume Cytokines like IL-1 IL-2 IL-4 IL-5 and IL-6 can be quantified The kit is optimized and validated for research use only
Catalog No. 50-000-80919 Supplier Meso Scale Discovery Supplier No. K15608U2
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Human Cytokine 30-plex Kit is an immunoassay for measuring 30 human cytokines and chemokines simultaneously It uses electrochemiluminescence detection on the MSD platform The kit includes a plate with capture antibodies detection antibodies and calibrator diluents The assay takes about 3 5 hours and works with serum plasma culture supernatants and other samples It offers high sensitivity a broad dynamic range and low sample volume Cytokines like IL-1 IL-2 IL-4 IL-5 and IL-6 can be quantified The kit is optimized and validated for research use only