Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 27 IgG Kit
SDP
Catalog No. 5000081465
Encompass
The V-PLEX SARS-CoV-2 Panel 2 IgG Kit from Meso Scale Diagnostics is an immunoassay that uses electrochemiluminescence technology to detect IgG antibodies against SARS-CoV-2 in human serum and plasma This qualitative assay can aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection The kit includes a 96-well plate coated with SARS-CoV-2 antigen detection antibodies calibrators controls diluent wash buffer read buffer plate seals and an MSD instrument for analysis Results are available in under 3 hours This assay has high sensitivity and specificity for detecting IgG antibodies
Catalog No. 50-000-81465 Supplier Meso Scale Discovery Supplier No. K15606U2
Add to Cart
Add to Cart
Request a Quote
The V-PLEX SARS-CoV-2 Panel 2 IgG Kit from Meso Scale Diagnostics is an immunoassay that uses electrochemiluminescence technology to detect IgG antibodies against SARS-CoV-2 in human serum and plasma This qualitative assay can aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection The kit includes a 96-well plate coated with SARS-CoV-2 antigen detection antibodies calibrators controls diluent wash buffer read buffer plate seals and an MSD instrument for analysis Results are available in under 3 hours This assay has high sensitivity and specificity for detecting IgG antibodies