Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 27 IgG
SDP
Catalog No. 5000078347
Encompass
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 2 IgG Kit is an immunoassay that quantitatively measures IgG antibodies to SARS-CoV-2 in human serum and plasma It uses electrochemiluminescence detection technology and provides high sensitivity specificity and reproducibility The kit includes a 96-well plate coated with SARS-CoV-2 spike protein S1 and receptor binding domain antigens detection antibodies calibrators controls wash buffer read buffer and plate seals Results are reported in arbitrary units per milliliter and the assay has a run time of approximately 3 hours This product helps monitor immune response to SARS-CoV-2
Catalog No. 50-000-78347 Supplier Meso Scale Discovery Supplier No. K15606U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 2 IgG Kit is an immunoassay that quantitatively measures IgG antibodies to SARS-CoV-2 in human serum and plasma It uses electrochemiluminescence detection technology and provides high sensitivity specificity and reproducibility The kit includes a 96-well plate coated with SARS-CoV-2 spike protein S1 and receptor binding domain antigens detection antibodies calibrators controls wash buffer read buffer and plate seals Results are reported in arbitrary units per milliliter and the assay has a run time of approximately 3 hours This product helps monitor immune response to SARS-CoV-2