Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 27 IgM Kit
SDP
Catalog No. 5000081187
Encompass
The Meso Scale Discovery COVID-19 Spike Protein Neutralization Assay Kit is a 96-well plate assay that measures neutralizing antibodies against SARS-CoV-2 in serum and plasma samples The kit uses Meso Scale Discovery electrochemiluminescence technology and includes SARS-CoV-2 Spike protein coated plates and calibrators to quantify antibody neutralization titers The assay has high sensitivity and dynamic range provides quantitative data and allows high throughput sample analysis It is intended for research use only to aid in understanding immune responses against SARS-CoV-2
Catalog No. 50-000-81187 Supplier Meso Scale Discovery Supplier No. K15607U2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery COVID-19 Spike Protein Neutralization Assay Kit is a 96-well plate assay that measures neutralizing antibodies against SARS-CoV-2 in serum and plasma samples The kit uses Meso Scale Discovery electrochemiluminescence technology and includes SARS-CoV-2 Spike protein coated plates and calibrators to quantify antibody neutralization titers The assay has high sensitivity and dynamic range provides quantitative data and allows high throughput sample analysis It is intended for research use only to aid in understanding immune responses against SARS-CoV-2