Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 27 MSE IgA
SDP
Catalog No. 5000080883
Encompass
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 2 Mouse IgA Kit is designed to quantitatively measure IgA antibodies specific to SARS-CoV-2 in mouse serum and plasma samples This electrochemiluminescence immunoassay allows high-throughput testing of samples for the presence of IgA antibodies binding to the receptor binding domain (RBD) of the SARS-CoV-2 spike protein The kit includes a plate coated with SARS-CoV-2 RBD antigen detection antibodies calibrators controls buffers and substrate Results are obtained in under 3 hours using an MSD instrument This assay can aid in evaluating vaccine efficacy and immunogenicity in mouse models of COVID-19
Catalog No. 50-000-80883 Supplier Meso Scale Discovery Supplier No. K15612U2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 2 Mouse IgA Kit is designed to quantitatively measure IgA antibodies specific to SARS-CoV-2 in mouse serum and plasma samples This electrochemiluminescence immunoassay allows high-throughput testing of samples for the presence of IgA antibodies binding to the receptor binding domain (RBD) of the SARS-CoV-2 spike protein The kit includes a plate coated with SARS-CoV-2 RBD antigen detection antibodies calibrators controls buffers and substrate Results are obtained in under 3 hours using an MSD instrument This assay can aid in evaluating vaccine efficacy and immunogenicity in mouse models of COVID-19