Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 27 MSE IgM
SDP
Catalog No. 5000080932
Encompass
The Mesoscale K15611U-4 is a 4-channel USB data acquisition module with 16-bit resolution 500 kS/s maximum sampling rate and 10 V input range It has 4 analog inputs 1 analog output 32 digital I/O and 2 counter/timers The module is bus-powered offers USB 2 0 compatibility and includes InstaCal software for Windows With its variety of I/O options high sampling rate and software for quick set-up the K15611U-4 is well-suited for portable data logging testing and measurement applications
Catalog No. 50-000-80932 Supplier Meso Scale Discovery Supplier No. K15611U4
Add to Cart
Add to Cart
Request a Quote
The Mesoscale K15611U-4 is a 4-channel USB data acquisition module with 16-bit resolution 500 kS/s maximum sampling rate and 10 V input range It has 4 analog inputs 1 analog output 32 digital I/O and 2 counter/timers The module is bus-powered offers USB 2 0 compatibility and includes InstaCal software for Windows With its variety of I/O options high sampling rate and software for quick set-up the K15611U-4 is well-suited for portable data logging testing and measurement applications