Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 28 ACE2
SDP
Catalog No. 5000079283
Encompass
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 2 (ACE2) Kit is a multiplex immunoassay that detects antibodies to SARS-CoV-2 in human serum and plasma It includes antigens for the receptor binding domain (RBD) of the SARS-CoV-2 spike protein and the ACE2 protein The assay has high sensitivity and specificity for detecting antibodies that block binding between the RBD and ACE2 which may have neutralizing activity against SARS-CoV-2 Results are obtained in just a few hours using an electrochemiluminescent detection method on Meso Scale Discovery s instrumentation platforms The kit includes reagents for 96 tests when used on MSD s SECTOR Imager 6000
Catalog No. 50-000-79283 Supplier Meso Scale Discovery Supplier No. K15617U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 2 (ACE2) Kit is a multiplex immunoassay that detects antibodies to SARS-CoV-2 in human serum and plasma It includes antigens for the receptor binding domain (RBD) of the SARS-CoV-2 spike protein and the ACE2 protein The assay has high sensitivity and specificity for detecting antibodies that block binding between the RBD and ACE2 which may have neutralizing activity against SARS-CoV-2 Results are obtained in just a few hours using an electrochemiluminescent detection method on Meso Scale Discovery s instrumentation platforms The kit includes reagents for 96 tests when used on MSD s SECTOR Imager 6000