Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 28 IgM Kit
SDP
Catalog No. 5000078605
Encompass
The Mesoscale K15615U-2 is a high-performance thermoelectric module for cooling heating and temperature control It has 127 thermocouples a max temperature differential of 70 C and measures 40 x 40 mm with a thickness of 3 4 mm With a Qmax of 15 1W and Imax of 6 2A it efficiently heats or cools various devices A heat sink and fan are needed for optimal performance Key features include high reliability RoHS compliance and a max operating temperature of 200 C
Catalog No. 50-000-78605 Supplier Meso Scale Discovery Supplier No. K15615U2
Add to Cart
Add to Cart
Request a Quote
The Mesoscale K15615U-2 is a high-performance thermoelectric module for cooling heating and temperature control It has 127 thermocouples a max temperature differential of 70 C and measures 40 x 40 mm with a thickness of 3 4 mm With a Qmax of 15 1W and Imax of 6 2A it efficiently heats or cools various devices A heat sink and fan are needed for optimal performance Key features include high reliability RoHS compliance and a max operating temperature of 200 C