Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 28 IgM
SDP
Catalog No. 5000081027
Encompass
The V-PLEX SARS-CoV-2 Panel 28 IgM Kit from Meso Scale Diagnostics is an immunoassay that detects IgM antibodies to SARS-CoV-2 in human serum and plasma This assay utilizes MSD s electrochemiluminescence technology and provides quantitative results for IgM antibodies to the receptor binding domain and nucleocapsid protein of SARS-CoV-2 The V-PLEX SARS-CoV-2 Panel 28 IgM Kit includes a 96-well plate coated with antigens detection antibodies calibrators and controls This assay can aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection
Catalog No. 50-000-81027 Supplier Meso Scale Discovery Supplier No. K15615U4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX SARS-CoV-2 Panel 28 IgM Kit from Meso Scale Diagnostics is an immunoassay that detects IgM antibodies to SARS-CoV-2 in human serum and plasma This assay utilizes MSD s electrochemiluminescence technology and provides quantitative results for IgM antibodies to the receptor binding domain and nucleocapsid protein of SARS-CoV-2 The V-PLEX SARS-CoV-2 Panel 28 IgM Kit includes a 96-well plate coated with antigens detection antibodies calibrators and controls This assay can aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection