Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 28 MSE IgA
SDP
Catalog No. 5000080965
Encompass
The Meso Scale Discovery K15620U-4 is a 96-well plate coated with a proprietary carbon electrode sensor that enables label-free detection of proteins and other biomolecules using electrochemiluminescence (ECL) It is designed for high-sensitivity quantitative analysis and multiplexing of up to 10 protein biomarkers per well The carbon electrode surface provides low background and high signal-to-noise ratio This product requires an MSD instrument such as the MESO QuickPlex SQ 120 for detection It has a 12 month shelf life when stored at 2-8 C
Catalog No. 50-000-80965 Supplier Meso Scale Discovery Supplier No. K15620U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery K15620U-4 is a 96-well plate coated with a proprietary carbon electrode sensor that enables label-free detection of proteins and other biomolecules using electrochemiluminescence (ECL) It is designed for high-sensitivity quantitative analysis and multiplexing of up to 10 protein biomarkers per well The carbon electrode surface provides low background and high signal-to-noise ratio This product requires an MSD instrument such as the MESO QuickPlex SQ 120 for detection It has a 12 month shelf life when stored at 2-8 C