Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 28 MSE IgG
SDP
Catalog No. 5000080933
Encompass
The Mesoscale K15618U-2 is a high-performance thermoelectric cooler that provides precise temperature control for photonics and semiconductor applications It features a maximum heat pumping capacity of 15 5W and can achieve a maximum temperature differential of 68 C This Peltier cooler has a low profile compact design with dimensions of 40 x 40 x 3 4mm It requires a supply voltage of 2 2VDC and can draw up to 6 5A of current With a weight of just 22 grams the K15618U-2 provides reliable solid-state heating and cooling in a small footprint
Catalog No. 50-000-80933 Supplier Meso Scale Discovery Supplier No. K15618U2
Add to Cart
Add to Cart
Request a Quote
The Mesoscale K15618U-2 is a high-performance thermoelectric cooler that provides precise temperature control for photonics and semiconductor applications It features a maximum heat pumping capacity of 15 5W and can achieve a maximum temperature differential of 68 C This Peltier cooler has a low profile compact design with dimensions of 40 x 40 x 3 4mm It requires a supply voltage of 2 2VDC and can draw up to 6 5A of current With a weight of just 22 grams the K15618U-2 provides reliable solid-state heating and cooling in a small footprint