Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 28 MSE IgM
SDP
Catalog No. 5000080787
Encompass
The Mesoscale K15619U-2 is a high-performance thermoelectric module that can be used for cooling heating and temperature control applications It consists of 127 thermocouples made from Bismuth Telluride semiconductor material sandwiched between two ceramic substrates Key features include a maximum temperature difference of 69 C an operating current of 3 4 Amps and dimensions of 40 x 40 x 3 4mm With a high coefficient of performance this Peltier module can provide efficient and precise temperature control for small spaces The K15619U-2 requires a heat sink and fan for heat dissipation
Catalog No. 50-000-80787 Supplier Meso Scale Discovery Supplier No. K15619U2
Add to Cart
Add to Cart
Request a Quote
The Mesoscale K15619U-2 is a high-performance thermoelectric module that can be used for cooling heating and temperature control applications It consists of 127 thermocouples made from Bismuth Telluride semiconductor material sandwiched between two ceramic substrates Key features include a maximum temperature difference of 69 C an operating current of 3 4 Amps and dimensions of 40 x 40 x 3 4mm With a high coefficient of performance this Peltier module can provide efficient and precise temperature control for small spaces The K15619U-2 requires a heat sink and fan for heat dissipation