Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 28 MSE IgM
SDP
Catalog No. 5000080788
Encompass
The V-PLEX SARS-CoV-2 Panel 28 (Mouse IgM) Kit (25 PL) from Meso Scale Diagnostics is a multiplex immunoassay kit for the qualitative detection of IgM antibodies to SARS-CoV-2 in mouse serum and plasma samples The kit contains a 96-well plate coated with antigens for SARS-CoV-2 calibrators controls detection antibody read buffer wash buffer and plate seals The assay utilizes electrochemiluminescence for detection and has a testing time of approximately 3 hours It is intended for research use only and not for diagnostic procedures The kit contains sufficient reagents for 25 plates which can test up to 700 samples when used as directed
Catalog No. 50-000-80788 Supplier Meso Scale Discovery Supplier No. K15619U4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX SARS-CoV-2 Panel 28 (Mouse IgM) Kit (25 PL) from Meso Scale Diagnostics is a multiplex immunoassay kit for the qualitative detection of IgM antibodies to SARS-CoV-2 in mouse serum and plasma samples The kit contains a 96-well plate coated with antigens for SARS-CoV-2 calibrators controls detection antibody read buffer wash buffer and plate seals The assay utilizes electrochemiluminescence for detection and has a testing time of approximately 3 hours It is intended for research use only and not for diagnostic procedures The kit contains sufficient reagents for 25 plates which can test up to 700 samples when used as directed