Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 29 IgM Kit
SDP
Catalog No. 5000081087
Encompass
The V-PLEX SARS-CoV-2 Panel 2 (IgM) Kit from Meso Scale Diagnostics is an immunoassay that detects IgM antibodies to SARS-CoV-2 in human serum and plasma This assay uses electrochemiluminescence technology on the MSD platform to quantitatively measure IgM antibodies specific to the SARS-CoV-2 nucleocapsid protein The kit includes a 96-well plate coated with SARS-CoV-2 nucleocapsid antigen detection antibody buffers calibrators and controls Results are available in under 3 hours from sample addition This assay aids in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection
Catalog No. 50-000-81087 Supplier Meso Scale Discovery Supplier No. K15625U2
Add to Cart
Add to Cart
Request a Quote
The V-PLEX SARS-CoV-2 Panel 2 (IgM) Kit from Meso Scale Diagnostics is an immunoassay that detects IgM antibodies to SARS-CoV-2 in human serum and plasma This assay uses electrochemiluminescence technology on the MSD platform to quantitatively measure IgM antibodies specific to the SARS-CoV-2 nucleocapsid protein The kit includes a 96-well plate coated with SARS-CoV-2 nucleocapsid antigen detection antibody buffers calibrators and controls Results are available in under 3 hours from sample addition This assay aids in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection