Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 29 MSE IgA
SDP
Catalog No. 5000080865
Encompass
The Mesoscale K15630U-4 is a 4-channel 30 MHz bandwidth 14-bit resolution digitizer with a maximum sampling rate of 250 MS/s It has a 1 GB on-board acquisition memory and features programmable input ranges DC offset adjustment and software-selectable AC/DC input coupling This PCIe digitizer card has a low noise floor and is well-suited for high-speed waveform capture and data acquisition applications The K15630U-4 utilizes a Xilinx Virtex-5 FPGA and Mesoscale s StreamStor technology for continuous streaming to host memory at rates up to 1 6 GB/s
Catalog No. 50-000-80865 Supplier Meso Scale Discovery Supplier No. K15630U4
Add to Cart
Add to Cart
Request a Quote
The Mesoscale K15630U-4 is a 4-channel 30 MHz bandwidth 14-bit resolution digitizer with a maximum sampling rate of 250 MS/s It has a 1 GB on-board acquisition memory and features programmable input ranges DC offset adjustment and software-selectable AC/DC input coupling This PCIe digitizer card has a low noise floor and is well-suited for high-speed waveform capture and data acquisition applications The K15630U-4 utilizes a Xilinx Virtex-5 FPGA and Mesoscale s StreamStor technology for continuous streaming to host memory at rates up to 1 6 GB/s