Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 29 MSE IgA
SDP
Catalog No. 5000081014
Encompass
The Mesoscale K15630U-2 is a high-performance thermoelectric cooler with a 30W heat pumping capacity It uses Bismuth Telluride semiconductor elements to transfer heat between ceramic plates when powered Features include a temperature difference over 70 C a 0-4A current range and a compact 30 x 30 x 3 4mm size Weighing 28 grams it offers efficient cooling A heat sink and fan are needed for thermal management Ideal for laser diode cooling medical diagnostics and temperature stabilization
Catalog No. 50-000-81014 Supplier Meso Scale Discovery Supplier No. K15630U2
Add to Cart
Add to Cart
Request a Quote
The Mesoscale K15630U-2 is a high-performance thermoelectric cooler with a 30W heat pumping capacity It uses Bismuth Telluride semiconductor elements to transfer heat between ceramic plates when powered Features include a temperature difference over 70 C a 0-4A current range and a compact 30 x 30 x 3 4mm size Weighing 28 grams it offers efficient cooling A heat sink and fan are needed for thermal management Ideal for laser diode cooling medical diagnostics and temperature stabilization