Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 29 MSE IgG
SDP
Catalog No. 5000079288
Encompass
The Mesoscale K15628U-4 is a high-performance thermoelectric cooler for photonics and semiconductor applications offering precise temperature control It has a heat pumping capacity of 15 5W and a temperature differential of 70 C With dimensions of 40 x 40 x 3 4mm it requires 15V and handles up to 6A The K15628U-4 uses a Bi2Te3 thermoelectric module and aluminum nitride ceramic substrate for optimal performance making it ideal for stabilizing laser diodes and cooling sensors
Catalog No. 50-000-79288 Supplier Meso Scale Discovery Supplier No. K15628U4
Add to Cart
Add to Cart
Request a Quote
The Mesoscale K15628U-4 is a high-performance thermoelectric cooler for photonics and semiconductor applications offering precise temperature control It has a heat pumping capacity of 15 5W and a temperature differential of 70 C With dimensions of 40 x 40 x 3 4mm it requires 15V and handles up to 6A The K15628U-4 uses a Bi2Te3 thermoelectric module and aluminum nitride ceramic substrate for optimal performance making it ideal for stabilizing laser diodes and cooling sensors