Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 29 MSE IgG
SDP
Catalog No. 5000080953
Encompass
The Meso Scale Discovery K15628U-2 is a 96-well plate coated with a proprietary carbon electrode sensor that enables label-free detection of proteins and other biomolecules using electrochemiluminescence (ECL) It is designed for high-sensitivity quantitative analysis and screening applications The carbon electrode sensor plates are compatible with MSD s SECTOR Imager instrument line and provide a customizable assay platform for protein biomarker research immunogenicity testing and other applications requiring sensitive multiplex protein measurement This product contains 2 plates allowing users to run up to 192 assays
Catalog No. 50-000-80953 Supplier Meso Scale Discovery Supplier No. K15628U2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery K15628U-2 is a 96-well plate coated with a proprietary carbon electrode sensor that enables label-free detection of proteins and other biomolecules using electrochemiluminescence (ECL) It is designed for high-sensitivity quantitative analysis and screening applications The carbon electrode sensor plates are compatible with MSD s SECTOR Imager instrument line and provide a customizable assay platform for protein biomarker research immunogenicity testing and other applications requiring sensitive multiplex protein measurement This product contains 2 plates allowing users to run up to 192 assays