Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 29 MSE IgM
SDP
Catalog No. 5000079299
Encompass
The Meso Scale Discovery SARS-CoV-2 Panel 29 Mouse IgM Kit is an immunoassay that detects IgM antibodies to SARS-CoV-2 in mouse serum and plasma samples The assay uses Meso Scale Discovery electrochemiluminescence technology and provides quantitative results in under 5 hours The panel allows for the measurement of antibodies specific to multiple SARS-CoV-2 antigens including the spike and nucleocapsid proteins This can aid in the evaluation of vaccine candidates and immunotherapies in mouse models of COVID-19 The kit includes a 96-well plate coated with antigens detection antibody calibrator and all other reagents needed to run the assay
Catalog No. 50-000-79299 Supplier Meso Scale Discovery Supplier No. K15629U2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery SARS-CoV-2 Panel 29 Mouse IgM Kit is an immunoassay that detects IgM antibodies to SARS-CoV-2 in mouse serum and plasma samples The assay uses Meso Scale Discovery electrochemiluminescence technology and provides quantitative results in under 5 hours The panel allows for the measurement of antibodies specific to multiple SARS-CoV-2 antigens including the spike and nucleocapsid proteins This can aid in the evaluation of vaccine candidates and immunotherapies in mouse models of COVID-19 The kit includes a 96-well plate coated with antigens detection antibody calibrator and all other reagents needed to run the assay