Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 30 IgA Kit
SDP
Catalog No. 5000078603
Encompass
The V-PLEX SARS-CoV-2 Panel 30 IgA Kit from Meso Scale Diagnostics is an immunoassay that uses electrochemiluminescence technology to detect IgA antibodies against SARS-CoV-2 in human serum and plasma This qualitative assay can aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection The kit includes a 96-well plate coated with SARS-CoV-2 antigen detection antibodies calibrators controls diluent wash buffer read buffer and plate seals Results are obtained in under 3 hours using Meso Scale Diagnostics instrumentation including the MESO QuickPlex SQ 120
Catalog No. 50-000-78603 Supplier Meso Scale Discovery Supplier No. K15634U2
Add to Cart
Add to Cart
Request a Quote
The V-PLEX SARS-CoV-2 Panel 30 IgA Kit from Meso Scale Diagnostics is an immunoassay that uses electrochemiluminescence technology to detect IgA antibodies against SARS-CoV-2 in human serum and plasma This qualitative assay can aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection The kit includes a 96-well plate coated with SARS-CoV-2 antigen detection antibodies calibrators controls diluent wash buffer read buffer and plate seals Results are obtained in under 3 hours using Meso Scale Diagnostics instrumentation including the MESO QuickPlex SQ 120