Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 30 IgM Kit
SDP
Catalog No. 5000080957
Encompass
The V-PLEX SARS-CoV-2 Panel 30 IgM Kit from Meso Scale Diagnostics is an immunoassay that uses electrochemiluminescence technology to detect IgM antibodies against SARS-CoV-2 in human serum and plasma This qualitative test can aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection The kit includes reagents for 30 tests and has a testing time of approximately 2 hours Results are analyzed using MSD reader systems This product can serve as an important tool in understanding immune response and seroprevalence
Catalog No. 50-000-80957 Supplier Meso Scale Discovery Supplier No. K15633U2
Add to Cart
Add to Cart
Request a Quote
The V-PLEX SARS-CoV-2 Panel 30 IgM Kit from Meso Scale Diagnostics is an immunoassay that uses electrochemiluminescence technology to detect IgM antibodies against SARS-CoV-2 in human serum and plasma This qualitative test can aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection The kit includes reagents for 30 tests and has a testing time of approximately 2 hours Results are analyzed using MSD reader systems This product can serve as an important tool in understanding immune response and seroprevalence