Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 30 MSE IgA
SDP
Catalog No. 5000080958
Encompass
The Mesoscale K15638U-2 is a high-performance thermoelectric cooler module for photonics and semiconductor applications It has a heat pumping capacity of 15 5W and a temperature differential of 68 C Measuring 15 8mm x 15 8mm x 3 4mm this Peltier cooler features a no load current of 2 7A and an open circuit voltage of 3 2V DC It uses bismuth telluride and a ceramic substrate for optimal thermal transfer Proper heat sinking and temperature control are needed for efficient operation making it ideal for stabilizing sensitive components
Catalog No. 50-000-80958 Supplier Meso Scale Discovery Supplier No. K15638U2
Add to Cart
Add to Cart
Request a Quote
The Mesoscale K15638U-2 is a high-performance thermoelectric cooler module for photonics and semiconductor applications It has a heat pumping capacity of 15 5W and a temperature differential of 68 C Measuring 15 8mm x 15 8mm x 3 4mm this Peltier cooler features a no load current of 2 7A and an open circuit voltage of 3 2V DC It uses bismuth telluride and a ceramic substrate for optimal thermal transfer Proper heat sinking and temperature control are needed for efficient operation making it ideal for stabilizing sensitive components