Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 31 ACE2
SDP
Catalog No. 5000078608
Encompass
The Meso Scale Discovery K15645U-4 is a 96-well plate coated with a proprietary carbon electrode sensor that enables label-free detection of biomolecules It is designed for quantitative measurement of total tau protein in human CSF and plasma samples using electrochemiluminescence (ECL) detection This assay kit includes a 96-well plate tau detection antibody sample diluent calibrator diluent wash buffer read buffer and calibrators to generate a standard curve The ECL signal is proportional to the amount of tau protein present in the sample This assay has high sensitivity and a broad dynamic range to quantitate total tau levels
Catalog No. 50-000-78608 Supplier Meso Scale Discovery Supplier No. K15645U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery K15645U-4 is a 96-well plate coated with a proprietary carbon electrode sensor that enables label-free detection of biomolecules It is designed for quantitative measurement of total tau protein in human CSF and plasma samples using electrochemiluminescence (ECL) detection This assay kit includes a 96-well plate tau detection antibody sample diluent calibrator diluent wash buffer read buffer and calibrators to generate a standard curve The ECL signal is proportional to the amount of tau protein present in the sample This assay has high sensitivity and a broad dynamic range to quantitate total tau levels