Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 31 IgA Kit
SDP
Catalog No. 5000080887
Encompass
The Meso Scale Discovery COVID-19 Antibody Discovery Kit is a research tool for detecting antibodies to SARS-CoV-2 in serum and plasma samples The electrochemiluminescence immunoassay uses SARS-CoV-2 spike protein coated plates and can quantitatively measure IgG IgM and IgA antibodies The kit includes coated plates detection antibody read buffer wash buffer sample diluent and calibrators to run the immunoassay The kit has high sensitivity and specificity for SARS-CoV-2 antibodies and is for research use only
Catalog No. 50-000-80887 Supplier Meso Scale Discovery Supplier No. K15644U2
The Meso Scale Discovery COVID-19 Antibody Discovery Kit is a research tool for detecting antibodies to SARS-CoV-2 in serum and plasma samples The electrochemiluminescence immunoassay uses SARS-CoV-2 spike protein coated plates and can quantitatively measure IgG IgM and IgA antibodies The kit includes coated plates detection antibody read buffer wash buffer sample diluent and calibrators to run the immunoassay The kit has high sensitivity and specificity for SARS-CoV-2 antibodies and is for research use only