Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 31 IgA
SDP
Catalog No. 5000080927
Encompass
The Meso Scale Discovery V-PLEX Human IL-15 Kit is a 4-plate assay kit that enables the quantitative measurement of human interleukin 15 (IL-15) in cell culture supernatants serum plasma and other biological fluids This electrochemiluminescent immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibody detection antibody conjugated to ruthenium calibrators and diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 2500 pg/mL
Catalog No. 50-000-80927 Supplier Meso Scale Discovery Supplier No. K15644U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Human IL-15 Kit is a 4-plate assay kit that enables the quantitative measurement of human interleukin 15 (IL-15) in cell culture supernatants serum plasma and other biological fluids This electrochemiluminescent immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibody detection antibody conjugated to ruthenium calibrators and diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 2500 pg/mL