Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 31 IgG Kit
SDP
Catalog No. 5000080938
Encompass
The Simoa SARS-CoV-2 N Protein Antigen Test is an ultrasensitive immunoassay that detects the nucleocapsid protein antigen of SARS-CoV-2 in serum plasma whole blood and nasal swab samples It uses Quanterix s Simoa technology which is up to 1000 times more sensitive than standard ELISA tests allowing earlier detection of infection The fully automated test runs on Quanterix s Simoa HD-X analyzer and provides quantitative results in 70 minutes This highly sensitive assay aids in diagnosis of acute infection and helps monitor disease progression With a limit of detection down to 0 01 pg/mL the Simoa SARS-CoV-2 N Protein Antigen Test offers industry-leading sensitivity
Catalog No. 50-000-80938 Supplier Meso Scale Discovery Supplier No. K15642U2
The Simoa SARS-CoV-2 N Protein Antigen Test is an ultrasensitive immunoassay that detects the nucleocapsid protein antigen of SARS-CoV-2 in serum plasma whole blood and nasal swab samples It uses Quanterix s Simoa technology which is up to 1000 times more sensitive than standard ELISA tests allowing earlier detection of infection The fully automated test runs on Quanterix s Simoa HD-X analyzer and provides quantitative results in 70 minutes This highly sensitive assay aids in diagnosis of acute infection and helps monitor disease progression With a limit of detection down to 0 01 pg/mL the Simoa SARS-CoV-2 N Protein Antigen Test offers industry-leading sensitivity