Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 31 MSE IgG
SDP
Catalog No. 5000081052
Encompass
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 31 Mouse IgG Kit is an electrochemiluminescence immunoassay for detecting IgG antibodies to SARS-CoV-2 in mouse serum and plasma It measures antibodies to the spike protein receptor binding domain and nucleocapsid protein using a multiplex format The kit includes a plate with antigens detection antibodies calibrators controls diluent read buffer and wash buffer The 3 5-hour protocol provides quantitative results for SARS-CoV-2 IgG levels enabling researchers to evaluate the humoral immune response and measure neutralizing antibodies in mouse models
Catalog No. 50-000-81052 Supplier Meso Scale Discovery Supplier No. K15646U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 31 Mouse IgG Kit is an electrochemiluminescence immunoassay for detecting IgG antibodies to SARS-CoV-2 in mouse serum and plasma It measures antibodies to the spike protein receptor binding domain and nucleocapsid protein using a multiplex format The kit includes a plate with antigens detection antibodies calibrators controls diluent read buffer and wash buffer The 3 5-hour protocol provides quantitative results for SARS-CoV-2 IgG levels enabling researchers to evaluate the humoral immune response and measure neutralizing antibodies in mouse models