Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 32 ACE2
SDP
Catalog No. 5000080888
Encompass
The Meso Scale Discovery SARS-CoV-2 Spike BA 5 Protein is a recombinant protein corresponding to the spike protein of the SARS-CoV-2 Omicron BA 5 variant It can be used in immunoassays and as an immunogen for antibody generation The protein is produced in HEK293 cells and contains a His-tag for purification It is supplied lyophilized from a Tris buffer The molecular weight is approximately 150 kDa for the trimeric protein This reagent allows researchers to study immune responses to the BA 5 variant of SARS-CoV-2
Catalog No. 50-000-80888 Supplier Meso Scale Discovery Supplier No. K15671U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery SARS-CoV-2 Spike BA 5 Protein is a recombinant protein corresponding to the spike protein of the SARS-CoV-2 Omicron BA 5 variant It can be used in immunoassays and as an immunogen for antibody generation The protein is produced in HEK293 cells and contains a His-tag for purification It is supplied lyophilized from a Tris buffer The molecular weight is approximately 150 kDa for the trimeric protein This reagent allows researchers to study immune responses to the BA 5 variant of SARS-CoV-2