Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 32 IgA Kit
SDP
Catalog No. 5000080921
Encompass
The V-PLEX SARS-CoV-2 Panel 2 IgA Kit from Meso Scale Diagnostics is an immunoassay that detects IgA antibodies to SARS-CoV-2 in human serum and plasma This assay uses electrochemiluminescence technology on the MSD platform to quantitatively measure IgA antibodies to the SARS-CoV-2 spike and nucleocapsid proteins The kit includes a 96-well plate coated with SARS-CoV-2 antigens detection antibodies calibrators controls diluent and other assay reagents required to perform the test according to the manufacturer s instructions Results are reported as index values relative to calibrators and controls This assay can aid in understanding the IgA antibody response to SARS-CoV-2 infection
Catalog No. 50-000-80921 Supplier Meso Scale Discovery Supplier No. K15670U2
Add to Cart
Add to Cart
Request a Quote
The V-PLEX SARS-CoV-2 Panel 2 IgA Kit from Meso Scale Diagnostics is an immunoassay that detects IgA antibodies to SARS-CoV-2 in human serum and plasma This assay uses electrochemiluminescence technology on the MSD platform to quantitatively measure IgA antibodies to the SARS-CoV-2 spike and nucleocapsid proteins The kit includes a 96-well plate coated with SARS-CoV-2 antigens detection antibodies calibrators controls diluent and other assay reagents required to perform the test according to the manufacturer s instructions Results are reported as index values relative to calibrators and controls This assay can aid in understanding the IgA antibody response to SARS-CoV-2 infection