Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 32 IgA
SDP
Catalog No. 5000080893
Encompass
The K15670U-4 is a 4-channel 16-bit 250 kS/s simultaneous sampling data acquisition module from Keithley Instruments It has 4 analog input channels with programmable input ranges of 10 V 5 V 2 V 1 V 500 mV 200 mV 100 mV and 50 mV The module has 16-bit resolution 250 kS/s maximum aggregate sample rate and includes screw terminal connectors It is compatible with the Keithley KPCI-3108 Series data acquisition cards The K15670U-4 is ideal for precision low-speed voltage measurement applications
Catalog No. 50-000-80893 Supplier Meso Scale Discovery Supplier No. K15670U4
Add to Cart
Add to Cart
Request a Quote
The K15670U-4 is a 4-channel 16-bit 250 kS/s simultaneous sampling data acquisition module from Keithley Instruments It has 4 analog input channels with programmable input ranges of 10 V 5 V 2 V 1 V 500 mV 200 mV 100 mV and 50 mV The module has 16-bit resolution 250 kS/s maximum aggregate sample rate and includes screw terminal connectors It is compatible with the Keithley KPCI-3108 Series data acquisition cards The K15670U-4 is ideal for precision low-speed voltage measurement applications