Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 32 IgG Kit
SDP
Catalog No. 5000079279
Encompass
The V-PLEX SARS-CoV-2 Panel 32 IgG Kit from Meso Scale Diagnostics is an immunoassay that quantitatively measures IgG antibodies to SARS-CoV-2 in human serum and plasma This kit includes a 96-well plate coated with SARS-CoV-2 spike protein antigens detection antibodies calibrators controls diluent wash buffer read buffer and plate seals The electrochemiluminescence immunoassay takes approximately 3 5 hours to run and has a dynamic range of 50-100 000 U/mL This assay can aid in understanding IgG seroprevalence and immune response to SARS-CoV-2
Catalog No. 50-000-79279 Supplier Meso Scale Discovery Supplier No. K15668U2
Add to Cart
Add to Cart
Request a Quote
The V-PLEX SARS-CoV-2 Panel 32 IgG Kit from Meso Scale Diagnostics is an immunoassay that quantitatively measures IgG antibodies to SARS-CoV-2 in human serum and plasma This kit includes a 96-well plate coated with SARS-CoV-2 spike protein antigens detection antibodies calibrators controls diluent wash buffer read buffer and plate seals The electrochemiluminescence immunoassay takes approximately 3 5 hours to run and has a dynamic range of 50-100 000 U/mL This assay can aid in understanding IgG seroprevalence and immune response to SARS-CoV-2