Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 32 IgG
SDP
Catalog No. 5000080884
Encompass
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 2 IgG Kit is an electrochemiluminescence immunoassay that quantitatively measures IgG antibodies to SARS-CoV-2 in human serum and plasma This assay uses a multiplex format to measure antibodies to the SARS-CoV-2 spike and nucleocapsid proteins The kit includes a calibrator to report quantitative results in Units/mL and has a dynamic range of 150-100 000 Units/mL This assay can aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection
Catalog No. 50-000-80884 Supplier Meso Scale Discovery Supplier No. K15668U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 2 IgG Kit is an electrochemiluminescence immunoassay that quantitatively measures IgG antibodies to SARS-CoV-2 in human serum and plasma This assay uses a multiplex format to measure antibodies to the SARS-CoV-2 spike and nucleocapsid proteins The kit includes a calibrator to report quantitative results in Units/mL and has a dynamic range of 150-100 000 Units/mL This assay can aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection