Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 32 IgM Kit
SDP
Catalog No. 5000080892
Encompass
The V-PLEX SARS-CoV-2 Panel 2 IgM Kit from Meso Scale Diagnostics is an immunoassay that detects IgM antibodies to SARS-CoV-2 in human serum and plasma This qualitative assay uses electrochemiluminescence technology and provides results in under 3 hours The kit includes plates coated with SARS-CoV-2 antigens detection antibodies calibrators controls wash buffers substrate and plastic consumables needed to run the assay This kit can aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection
Catalog No. 50-000-80892 Supplier Meso Scale Discovery Supplier No. K15669U2
Add to Cart
Add to Cart
Request a Quote
The V-PLEX SARS-CoV-2 Panel 2 IgM Kit from Meso Scale Diagnostics is an immunoassay that detects IgM antibodies to SARS-CoV-2 in human serum and plasma This qualitative assay uses electrochemiluminescence technology and provides results in under 3 hours The kit includes plates coated with SARS-CoV-2 antigens detection antibodies calibrators controls wash buffers substrate and plastic consumables needed to run the assay This kit can aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection