Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 32 IgM
SDP
Catalog No. 5000079278
Encompass
The Mesoscale K15669U-4 is a 4-channel 16-bit 250 kS/s simultaneous sampling data acquisition module It has 4 analog voltage input channels with programmable input ranges of 10 V 5 V 2 V and 1 V The module has 16-bit resolution and a maximum sampling rate of 250 kS/s It can perform 4-channel simultaneous sampling and streaming to PC memory through USB 2 0 The K15669U-4 has software-selectable AC/DC coupling and input impedances of 1 M and 50 It is compatible with both Windows and Linux operating systems The module is bus-powered and compact in size
Catalog No. 50-000-79278 Supplier Meso Scale Discovery Supplier No. K15669U4
Add to Cart
Add to Cart
Request a Quote
The Mesoscale K15669U-4 is a 4-channel 16-bit 250 kS/s simultaneous sampling data acquisition module It has 4 analog voltage input channels with programmable input ranges of 10 V 5 V 2 V and 1 V The module has 16-bit resolution and a maximum sampling rate of 250 kS/s It can perform 4-channel simultaneous sampling and streaming to PC memory through USB 2 0 The K15669U-4 has software-selectable AC/DC coupling and input impedances of 1 M and 50 It is compatible with both Windows and Linux operating systems The module is bus-powered and compact in size