Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 32 MSE IgG
SDP
Catalog No. 5000078601
Encompass
The Mesoscale K15672U-2 is a high-performance thermoelectric module for cooling featuring 127 Bismuth Telluride thermocouples between ceramic substrates It achieves a 70 C temperature difference and 15 4 Watts cooling power Measuring 40 x 40 x 3 4mm it has a low profile for effective heat spreading Optimal operation is at 25 C hot side and 0 C cold side requiring a heat sink and cooling device Ideal for laser diode cooling medical diagnostics and scientific instrumentation
Catalog No. 50-000-78601 Supplier Meso Scale Discovery Supplier No. K15672U2
Add to Cart
Add to Cart
Request a Quote
The Mesoscale K15672U-2 is a high-performance thermoelectric module for cooling featuring 127 Bismuth Telluride thermocouples between ceramic substrates It achieves a 70 C temperature difference and 15 4 Watts cooling power Measuring 40 x 40 x 3 4mm it has a low profile for effective heat spreading Optimal operation is at 25 C hot side and 0 C cold side requiring a heat sink and cooling device Ideal for laser diode cooling medical diagnostics and scientific instrumentation