Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 32 MSE IgG
SDP
Catalog No. 5000078602
Encompass
The Mesoscale K15672U-4 is a 4-channel 16-bit 250 kS/s simultaneous sampling data acquisition module It has 4 analog voltage input channels with programmable gain amplifiers anti-aliasing filters and 16-bit ADCs The input voltage range is 10 V with software-selectable gains of 1 2 4 8 It has a USB 2 0 interface and is compatible with Windows Linux and Mac operating systems The module is bus-powered and compact in size It is well suited for low-frequency signal acquisition applications
Catalog No. 50-000-78602 Supplier Meso Scale Discovery Supplier No. K15672U4
Add to Cart
Add to Cart
Request a Quote
The Mesoscale K15672U-4 is a 4-channel 16-bit 250 kS/s simultaneous sampling data acquisition module It has 4 analog voltage input channels with programmable gain amplifiers anti-aliasing filters and 16-bit ADCs The input voltage range is 10 V with software-selectable gains of 1 2 4 8 It has a USB 2 0 interface and is compatible with Windows Linux and Mac operating systems The module is bus-powered and compact in size It is well suited for low-frequency signal acquisition applications