Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 32 MSE IgM
SDP
Catalog No. 5000080885
Encompass
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 4 Mouse IgM Kit is an electrochemiluminescence immunoassay that detects IgM antibodies to 4 SARS-CoV-2 antigens in mouse serum and plasma samples The kit includes a 4-spot MULTI-ARRAY 96-well plate coated with Spike S1 protein Spike S2 protein Nucleocapsid protein and Receptor Binding Domain along with detection antibodies calibrators controls diluent read buffer and wash buffer sufficient for 25 tests in a 32-well format The assay has a testing time of approximately 3 hours and 10 minutes and a sample volume requirement of 25 L
Catalog No. 50-000-80885 Supplier Meso Scale Discovery Supplier No. K15673U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 4 Mouse IgM Kit is an electrochemiluminescence immunoassay that detects IgM antibodies to 4 SARS-CoV-2 antigens in mouse serum and plasma samples The kit includes a 4-spot MULTI-ARRAY 96-well plate coated with Spike S1 protein Spike S2 protein Nucleocapsid protein and Receptor Binding Domain along with detection antibodies calibrators controls diluent read buffer and wash buffer sufficient for 25 tests in a 32-well format The assay has a testing time of approximately 3 hours and 10 minutes and a sample volume requirement of 25 L