Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 33 ACE2 Kit
SDP
Catalog No. 5000081050
Encompass
The V-PLEX SARS-CoV-2 Panel 33 Kit by Meso Scale Diagnostics is a serology test that detects antibodies to SARS-CoV-2 in serum and plasma samples It uses electrochemiluminescence technology and provides quantitative results for IgG antibodies to the spike and nucleocapsid proteins The test has high sensitivity and specificity and has received FDA Emergency Use Authorization It requires the MESO QuickPlex SQ 120 instrument and takes approximately 3 hours to run The kit includes a plate coated with antigens detection antibodies calibrators controls diluent and wash buffer It is intended for use in clinical labs and requires venipuncture whole blood samples
Catalog No. 50-000-81050 Supplier Meso Scale Discovery Supplier No. K15679U2
Add to Cart
Add to Cart
Request a Quote
The V-PLEX SARS-CoV-2 Panel 33 Kit by Meso Scale Diagnostics is a serology test that detects antibodies to SARS-CoV-2 in serum and plasma samples It uses electrochemiluminescence technology and provides quantitative results for IgG antibodies to the spike and nucleocapsid proteins The test has high sensitivity and specificity and has received FDA Emergency Use Authorization It requires the MESO QuickPlex SQ 120 instrument and takes approximately 3 hours to run The kit includes a plate coated with antigens detection antibodies calibrators controls diluent and wash buffer It is intended for use in clinical labs and requires venipuncture whole blood samples