Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 33 ACE2
SDP
Catalog No. 5000080955
Encompass
The V-PLEX SARS-CoV-2 Panel 33 Kit by Meso Scale Diagnostics is a serology test that detects antibodies to SARS-CoV-2 in serum and plasma samples It uses electrochemiluminescence technology and provides quantitative results for IgG antibodies to the spike and nucleocapsid proteins The panel allows for testing of up to 33 samples in duplicate on a single 96-well plate Results are available in as little as 3 hours This product helps track immune response and exposure to SARS-CoV-2
Catalog No. 50-000-80955 Supplier Meso Scale Discovery Supplier No. K15679U4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX SARS-CoV-2 Panel 33 Kit by Meso Scale Diagnostics is a serology test that detects antibodies to SARS-CoV-2 in serum and plasma samples It uses electrochemiluminescence technology and provides quantitative results for IgG antibodies to the spike and nucleocapsid proteins The panel allows for testing of up to 33 samples in duplicate on a single 96-well plate Results are available in as little as 3 hours This product helps track immune response and exposure to SARS-CoV-2