Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 33 IgA Kit
SDP
Catalog No. 5000080944
Encompass
The Mesoscale K15678U-2 is a high-performance thermoelectric module that can be used for cooling or heating applications It measures 40 x 40 x 3 4mm and provides a maximum heat pumping capacity of 15 8W The module operates on 15V and can achieve a maximum temperature difference of 68 C It utilizes Bismuth Telluride-based alloys as n-type and p-type semiconductor materials The module has a no-load voltage of 3 78V and internal resistance of 1 15 ohms It can withstand peak operating temperatures between -40 C to 80 C The K15678U-2 module is designed for regulated low noise thermoelectric applications
Catalog No. 50-000-80944 Supplier Meso Scale Discovery Supplier No. K15678U2
Add to Cart
Add to Cart
Request a Quote
The Mesoscale K15678U-2 is a high-performance thermoelectric module that can be used for cooling or heating applications It measures 40 x 40 x 3 4mm and provides a maximum heat pumping capacity of 15 8W The module operates on 15V and can achieve a maximum temperature difference of 68 C It utilizes Bismuth Telluride-based alloys as n-type and p-type semiconductor materials The module has a no-load voltage of 3 78V and internal resistance of 1 15 ohms It can withstand peak operating temperatures between -40 C to 80 C The K15678U-2 module is designed for regulated low noise thermoelectric applications